Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PMVK/phosphomevalonate kinase Antibody (21C10), Alexa Fluor™ 350, Novus Biologicals™
SDP

Catalog No. NB160719 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB160719 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB160719 Supplier Novus Biologicals Supplier No. NBP259412AF350
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

PMVK/phosphomevalonate kinase Monoclonal antibody specifically detects PMVK/phosphomevalonate kinase in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunofluorescence

Specifications

Antigen PMVK/phosphomevalonate kinase
Applications Western Blot, Flow Cytometry, ELISA, Immunofluorescence
Classification Monoclonal
Clone 21C10
Conjugate Alexa Fluor 350
Formulation 50mM Sodium Borate
Gene Alias hPMK, HUMPMKI, phosphomevalonate kinase, PMKA, PMKase, PMKEC 2.7.4.2, PMKI
Host Species Mouse
Immunogen Recombinant human PMVK/phosphomevalonate kinase (1-192aa) purified from E. coli
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, metabolism
Primary or Secondary Primary
Gene ID (Entrez) 10654
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.