Learn More
PMVK/phosphomevalonate kinase Antibody, DyLight 680, Novus Biologicals™
Shop All R&D Systems Products

Description
Specifications
Specifications
| Antigen | PMVK/phosphomevalonate kinase |
| Applications | ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | DyLight 680 |
| Dilution | ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin |
| Formulation | 50mM Sodium Borate |
| Gene Alias | hPMK, HUMPMKI, phosphomevalonate kinase, PMKA, PMKase, PMKEC 2.7.4.2, PMKI |
| Host Species | Rabbit |
| Immunogen | Produced in rabbits immunized with purified, recombinant PMVK/phosphomevalonate kinase (Q15126; Met1-Leu192). |
| Purification Method | Affinity purified |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.