Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PNCK Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP186652
Description
PNCK Polyclonal specifically detects PNCK in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
PNCK | |
Polyclonal | |
Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml, Immunohistochemistry-Paraffin 1:20-1:50 | |
BSTK3, calcium/calmodulin-dependent protein kinase type 1B, CaM kinase I beta, CaM kinase IB, CaMK1b, CaM-KI beta, CaMKI-beta, EC 2.7.11, EC 2.7.11.17, FLJ50403, FLJ50549, FLJ56451, FLJ59811, MGC45419, pregnancy up-regulated non-ubiquitously expressed CaM kinase, pregnancy upregulated non-ubiquitously expressed CaM kinase, Pregnancy up-regulated non-ubiquitously-expressed CaM kinase | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Unconjugated | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
PNCK | |
This antibody was developed against Recombinant Protein corresponding to amino acids:MLLLKKHTEDISSVYEIRERLGSGAFSEVVLAQERGSAHLVALKCIPKKALRGKEALVENEIAVLRRISHPN | |
0.1 mL | |
Protein Kinase | |
139728 | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction