Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Pneumolysin Mouse anti-Bacteria, Clone: 1F11, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP105134
Description
Pneumolysin Monoclonal specifically detects Pneumolysin in Bacteria samples. It is validated for Western Blot, ELISA.Specifications
Pneumolysin | |
Monoclonal | |
1.0 mg/mL | |
Western Blot 1:100-1:2000, ELISA 1:100-1:2000 | |
Pneumolysin from at least 9 different types of pneumococcal strains | |
100 μg | |
Primary | |
Bacteria | |
Purified |
Western Blot, ELISA | |
1F11 | |
Unconjugated | |
Mouse | |
Protein A or G purified | |
RUO | |
Reacts specifically with Pneumolysin from at least 9 different types of pneumococcal strains when tested by SDS-PAGE and WB. The tested pneumococcal types were 5, 6A, 11A, 11F, 12B, 15F, 19A, 25F and 35A. | |
Store at 4C. Do not freeze. | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction