Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Pneumolysin Mouse anti-Bacteria, Clone: 1F11, Novus Biologicals™

Mouse Monoclonal Antibody
Specifications
Antigen | Pneumolysin |
---|---|
Clone | 1F11 |
Concentration | 1.0 mg/mL |
Dilution | Western Blot 1:100-1:2000, ELISA 1:100-1:2000 |
Applications | Western Blot, ELISA |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP105134
![]() |
Novus Biologicals
NBP105134 |
100 μg | N/A | N/A | N/A | ||||
Description
Pneumolysin Monoclonal specifically detects Pneumolysin in Bacteria samples. It is validated for Western Blot, ELISA.Specifications
Pneumolysin | |
1.0 mg/mL | |
Western Blot, ELISA | |
Unconjugated | |
Mouse | |
Bacteria | |
IgG1 κ | |
Protein A or G purified | |
Reacts specifically with Pneumolysin from at least 9 different types of pneumococcal strains when tested by SDS-PAGE and WB. The tested pneumococcal types were 5, 6A, 11A, 11F, 12B, 15F, 19A, 25F and 35A. |
1F11 | |
Western Blot 1:100-1:2000, ELISA 1:100-1:2000 | |
Monoclonal | |
Purified | |
RUO | |
Pneumolysin from at least 9 different types of pneumococcal strains | |
Primary | |
Store at 4C. Do not freeze. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title