Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Promega PNGase F Recombinant Protein
Shop All Promega Corporation Products
Click to view available options
Quantity:
500 U
Description
- Recombinant Glycosidase Cloned from Elizabethkingia miricola
- Use to determine protein glycosylation status and location
- Cleaves N-linked oligosaccharides between innermost GlcNAc and asparagines from N-linked glycoproteins
- Will not remove oligosaccharides found on plant glycoproteins
- Supplied at a concentration of 10u/μL
Specifications
Specifications
| Concentration | 10U/μL |
| For Use With (Application) | Glycan Structure Characterization, Protein Glycosylation, Protein Trafficking |
| Molecular Weight (g/mol) | 36kDa |
| Quantity | 500 U |
| Source | Elizabethkingia miricola |
| Storage Requirements | 2 to 10°C |
| Recombinant | Recombinant |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction