Learn More
Promega pNL3.2.CMV Vector
Catalog No. prn1411 Shop All Promega Corporation Products
Click to view available options
Quantity:
20 μg
Catalog No. PRN1411
Supplier: Promega N1411
Only null left
Add to Cart
Add to Cart
NanoLuc (Nluc) luciferase is a small bright luminescent reporter enzyme (19.1kDa) engineered for optimal performance. Use the pNL3.2.CMV Vector as a negative control in experiments measuring regulated changes in NanoLuc luciferase expression.
  • Vector that Expresses NanoLuc(R) Luciferase Using the CMV Promoter
  • Used as a control vector for the NanoLuc(R) fusion vectors
  • Includes an optimized NanoLuc(R) luciferase gene that codes for a reporter protein with a shorter half-life (NlucP)
  • Hygromycin resistance selectable marker means choosing transient transfection or generating a stable cell line
Specifications
Quantity 20 μg
Content And Storage -30°C to -10°C
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.