Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PNMA3 Antibody (OTI1C3), PE/Cy7, Novus Biologicals™
SDP

Catalog No. N273485PEC7 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N273485PEC7 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N273485PEC7 Supplier Novus Biologicals Supplier No. NBP273485PECY7
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

PNMA3 Monoclonal specifically detects PNMA3 in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen PNMA3
Applications Flow Cytometry
Classification Monoclonal
Clone OTI1C3
Conjugate PE-Cyanine7
Dilution Flow Cytometry
Formulation PBS with 0.05% Sodium Azide
Gene Alias MA3MA5, MGC132756, MGC132758, paraneoplastic antigen MA3, paraneoplastic cancer-testis-brain antigen
Gene Symbols PNMA3
Host Species Mouse
Immunogen Full length recombinant protein of human PNMA3 (NP_037496) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 29944.0
Target Species Human
Content And Storage Store at 4°C in the dark. Do not freeze.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.