Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PNMT Antibody (CL11434), Novus Biologicals™
SDP

Catalog No. NB238063 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB238063 25 μg
NB236759 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB238063 Supplier Novus Biologicals Supplier No. NBP32119325UL
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

PNMT Monoclonal antibody specifically detects PNMT in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)

Specifications

Antigen PNMT
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone CL11434
Conjugate Unconjugated
Dilution Immunohistochemistry 1:200-1:500, Immunohistochemistry-Paraffin 1:200-1:500
Formulation PBS, pH 7.2, 40% glycerol
Gene Alias EC 2.1.1, EC 2.1.1.28, Noradrenaline N-methyltransferase, PENTMGC34570, phenylethanolamine N-methylase, phenylethanolamine N-methyltransferase, PNMTase
Host Species Mouse
Immunogen This antibody was developed against a Recombinant Protein Epitope Signature Tag (PrEST) antigen sequence of P11086. Actual immunogen sequence is proprietary.
Purification Method Protein A purified
Quantity 25 μg
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 5409
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.