Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PNPLA3 Mouse anti-Human, Unconjugated, Clone: 11C5, MilliporeSigma™

Mouse Monoclonal Antibody

Supplier:  MilliporeSigma MABS217425UG

 View more versions of this product

Catalog No. MABS21742MI


Only null left
Add to Cart

Description

Description

PNPLA3 Monoclonal antibody specifically detects PNPLA3 in Human samples. It is validated for Western Blotting.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

PNPLA3
Monoclonal
Unconjugated
1-acylglycerol-3-phosphate O-acyltransferase PNPLA3;EC:2.3.1.51;Acylglycerol transacylase;Adiponutrin;ADPN;Calcium-independent phospholipase A2-epsilon;iPLA2-epsilon;Lysophosphatidic acid acyltransferase;Patatin-like phospholipase domain-containing protein 3
Mouse
∼54 kDa observed; 52.86 kDa calculated. Uncharacterized bands may be observed in some lysate(s).
25 μg
Primary
Human
Purified
Western Blot
11C5
NP_079501
PNPLA3, ASPN1, C22irf20
Recombinant fusion protein of human PNPLA3 and Trigger Factor.
Protein G Purified
Signaling
Clone 11C5 is a mouse monoclonal antibody that detects human Patatin-like phospholipase domain-containing protein 3 (PNPLA3).
Stable for 1 year at 2 to 8°C from date of receipt.
IgG2a κ
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.