Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PNPLA3 Mouse anti-Human, Unconjugated, Clone: 11C5, MilliporeSigma™
Mouse Monoclonal Antibody
$188.90 - $478.50
Specifications
Antigen | PNPLA3 |
---|---|
Clone | 11C5 |
Applications | Western Blot |
Classification | Monoclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MABS21742MI
|
MilliporeSigma
MABS217425UG |
25 μg |
Each for $188.90
|
|
|||||
MABS21741MI
|
MilliporeSigma
MABS2174100UG |
100 μg |
Each for $478.50
|
|
|||||
Description
PNPLA3 Monoclonal antibody specifically detects PNPLA3 in Human samples. It is validated for Western Blotting.Specifications
PNPLA3 | |
Western Blot | |
Unconjugated | |
Mouse | |
Human | |
1-acylglycerol-3-phosphate O-acyltransferase PNPLA3;EC:2.3.1.51;Acylglycerol transacylase;Adiponutrin;ADPN;Calcium-independent phospholipase A2-epsilon;iPLA2-epsilon;Lysophosphatidic acid acyltransferase;Patatin-like phospholipase domain-containing protein 3 | |
Recombinant fusion protein of human PNPLA3 and Trigger Factor. | |
Primary | |
Stable for 1 year at 2 to 8°C from date of receipt. | |
∼54 kDa observed; 52.86 kDa calculated. Uncharacterized bands may be observed in some lysate(s). |
11C5 | |
Monoclonal | |
Purified | |
Signaling | |
NP_079501 | |
PNPLA3, ASPN1, C22irf20 | |
IgG2a κ | |
Protein G Purified | |
Clone 11C5 is a mouse monoclonal antibody that detects human Patatin-like phospholipase domain-containing protein 3 (PNPLA3). |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title