Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PNPLA3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5113494
Change view
Click to view available options
Quantity:
100 μL
Catalog No. Quantity
PIPA5113494 100 μL
1 options

Catalog No. PIPA5113494

Supplier: Invitrogen™ PA5113494

Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PNPLA3 Polyclonal Antibody for Western Blot, IHC (P), ELISA

The protein encoded by this gene is a triacylglycerol lipase that mediates triacylglycerol hydrolysis in adipocytes. The encoded protein, which appears to be membrane bound, may be involved in the balance of energy usage/storage in adipocytes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PNPLA3
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 3.95 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene PNPLA3
Gene Accession No. Q9NST1
Gene Alias 1-acylglycerol-3-phosphate O-acyltransferase Pnpla3; acylglycerol O-acyltransferase; Acylglycerol transacylase; Adiponutrin; ADPN; C22orf20; Calcium-independent phospholipase A2-epsilon; iPLA(2)epsilon; iPLA2epsilon; iPLA2-epsilon; Lysophosphatidic acid acyltransferase; patatin like phospholipase domain containing 3; patatin-like phospholipase domain containing 3; patatin-like phospholipase domain-containing protein 3; Pnpla3
Gene Symbols PNPLA3
Host Species Rabbit
Immunogen Recombinant Human Patatin-like phospholipase domain-containing protein 3 protein (377-481AA).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 80339
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.