Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PNPLA5 Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

$480.74

Specifications

Antigen PNPLA5
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Form Purified
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP170679
SDP
View Documents
Novus Biologicals
NBP170679
100 μL
Each for $480.74
Only null left
Add to Cart
 
NBP17067920
SDP
View Documents
Novus Biologicals
NBP17067920UL
20 μL N/A N/A N/A
Description

Description

PNPLA5 Polyclonal specifically detects PNPLA5 in Human samples. It is validated for Western Blot.
Specifications

Specifications

PNPLA5
Polyclonal
Purified
RUO
dJ388M5, dJ388M5.4, GS2L, patatin-like phospholipase domain containing 5, patatin-like phospholipase domain-containing protein 5
PNPLA5
IgG
Protein A purified
Western Blot
Unconjugated
Rabbit
Human
150379
Synthetic peptides corresponding to PNPLA5(patatin-like phospholipase domain containing 5) The peptide sequence was selected from the N terminal of PNPLA5. Peptide sequence LSLSILHPAYAPIEHVKQQLQDALPPDAHVLASQRLGISLTRWPDGRNFL.
Primary
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.