Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PNPO Antibody (OTI1H9), Novus Biologicals™
SDP

Catalog No. NBP201088 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP201088 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP201088 Supplier Novus Biologicals Supplier No. NBP201088
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

PNPO Monoclonal specifically detects PNPO in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen PNPO
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone OTI1H9
Concentration 0.32 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:150
Gene Alias EC 1.4.3.5, FLJ10535, PDXPO, pyridoxal 5'-phosphate synthase, pyridoxamine 5'-phosphate oxidase, Pyridoxamine-phosphate oxidase, pyridoxine 5'-phosphate oxidase, pyridoxine-5'-phosphate oxidase
Gene Symbols PNPO
Host Species Mouse
Immunogen Full length human recombinant protein of human PNPO(NP_060599) produced in HEK293T cell.
Molecular Weight of Antigen 29.8 kDa
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, metabolism
Primary or Secondary Primary
Gene ID (Entrez) 55163
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Isotype IgG2b
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.