Learn More
Description
Specifications
Specifications
| Antigen | PNPT1 |
| Applications | Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Janelia Fluor 549 |
| Dilution | Immunocytochemistry/Immunofluorescence |
| Formulation | 50mM Sodium Borate |
| Gene Alias | DKFZp762K1914, EC 2.7.7, EC 2.7.7.8, mitochondrial, old-35, OLD35, PNPase, PNPase 1, PNPase old-35, Polynucleotide phosphorylase 1, Polynucleotide phosphorylase-like protein, polyribonucleotide nucleotidyltransferase 1 |
| Host Species | Rabbit |
| Immunogen | Produced in rabbits immunized with E. coli-derived Human PNPT1 fragment. |
| Purification Method | Antigen Affinity-purified |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
