Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PNR/NR2E3 Antibody (OTI3D4), Alexa Fluor™ 488, Novus Biologicals™
SDP

Catalog No. NB273510A48 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB273510A48 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB273510A48 Supplier Novus Biologicals Supplier No. NBP273510AF488
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

PNR/NR2E3 Monoclonal specifically detects PNR/NR2E3 in Human, Mouse, Rat samples. It is validated for Western Blot.

Specifications

Antigen PNR/NR2E3
Applications Western Blot
Classification Monoclonal
Clone OTI3D4
Conjugate Alexa Fluor 488
Dilution Western Blot
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias Nuclear receptor subfamily 2 group E member 3, nuclear receptor subfamily 2, group E, member 3, photoreceptor-specific nuclear receptor, PNRESCS, rd7, Retina-specific nuclear receptor, RNR, RP37MGC49976
Gene Symbols NR2E3
Host Species Mouse
Immunogen Full length recombinant protein of human PNR/NR2E3 (NP_057430) produced in HEK293T cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline GPCR, Vision
Primary or Secondary Primary
Gene ID (Entrez) 10002.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.