Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Podocalyxin Like Antibody (192703), PE/Cy7, Novus Biologicals™
SDP

Catalog No. NB314405 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB314405 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB314405 Supplier Novus Biologicals Supplier No. FAB1556PECY7
Add to Cart
Add to Cart

Rat Monoclonal Antibody

Podocalyxin Like Monoclonal specifically detects Podocalyxin Like in Mouse samples. It is validated for Flow Cytometry.

Specifications

Antigen Podocalyxin Like
Applications Flow Cytometry
Classification Monoclonal
Clone 192703
Conjugate PE-Cyanine7
Dilution Flow Cytometry
Formulation PBS
Gene Alias Gp200MGC138240, PCLP1, PCLP-1, PCLPGCTM-2 antigen, PCpodocalyxin, podocalyxin-like, Podocalyxin-like protein 1
Host Species Rat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse Podocalyxin, Ser21-Arg402, Accession # Q9R0M4
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Cell Biology, Cellular Markers, Hematopoietic Stem Cell Markers, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 5420
Target Species Mouse
Content And Storage Store at 4C in the dark. Do not freeze.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.