Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Podocalyxin Like Antibody (3D3), PE/Cy7, Novus Biologicals™
SDP

Catalog No. N233108PEC7 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N233108PEC7 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N233108PEC7 Supplier Novus Biologicals Supplier No. NBP233108PECY7
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Podocalyxin Like Monoclonal specifically detects Podocalyxin Like in Human, Mouse, Rat, Rabbit samples. It is validated for Flow Cytometry.

Specifications

Antigen Podocalyxin Like
Applications Flow Cytometry
Classification Monoclonal
Clone 3D3
Conjugate PE-Cyanine7
Dilution Flow Cytometry
Formulation PBS with 0.05% Sodium Azide
Gene Alias Gp200MGC138240, PCLP1, PCLP-1, PCLPGCTM-2 antigen, PCpodocalyxin, podocalyxin-like, Podocalyxin-like protein 1
Gene Symbols PODXL
Host Species Mouse
Immunogen A recombinant protein fragment containing the intracellular, transmembrane, and part of the extracellular domain of human Podocalyxin Like (Uniprot: O00592)
Purification Method Protein G purified
Quantity 0.1 mL
Research Discipline Cancer, Cell Biology, Cellular Markers, Embryonic Stem Cell Markers, Hematopoietic Stem Cell Markers, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 5420.0
Test Specificity Podocalyxin is a member of the CD34 transmembrane sialomucin family. It is over-expressed on the podocyte foot projections and plays essential roles in kidney development and homeostasis, blood filtration and urine formation. It is also expressed on vascular endothelia, hematopoietic progenitors and a subset of neurons. Overexpression of podocalyxin may be linked to more aggressive tumor behavior. Podocalyxin antibody can identify podocytes in the urine (podocyturia) that may indicate glomerular disease, pre-eclampsia, and other kidney pathology.
Target Species Human, Mouse, Rat, Rabbit
Content And Storage Store at 4°C in the dark. Do not freeze.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.