Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Podocalyxin Like Antibody (OTI2C6), DyLight 488, Novus Biologicals™
SDP

Catalog No. NBP273511G Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP273511G 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP273511G Supplier Novus Biologicals Supplier No. NBP273511G
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Podocalyxin Like Monoclonal specifically detects Podocalyxin Like in Human samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen Podocalyxin Like
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone OTI2C6
Conjugate DyLight 488
Dilution Western Blot, Immunohistochemistry
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias Gp200MGC138240, PCLP1, PCLP-1, PCLPGCTM-2 antigen, PCpodocalyxin, podocalyxin-like, Podocalyxin-like protein 1
Gene Symbols PODXL
Host Species Mouse
Immunogen Recombinant protein fragment corresponding to amino acids 32-461 of human Podocalyxin Like (NP_005388) produced in HEK293 cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Cancer, Cell Biology, Cellular Markers, Embryonic Stem Cell Markers, Hematopoietic Stem Cell Markers, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 5420.0
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.