Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Podocin Monoclonal Antibody (C3), Invitrogen™
Mouse Monoclonal Antibody
$213.50 - $522.50
Specifications
Antigen | Podocin |
---|---|
Clone | C3 |
Concentration | 1 mg/mL |
Applications | Western Blot |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
PIMA543646
|
Invitrogen™
MA543646 |
20 μL |
Each for $213.50
|
|
|||||
PIMA543645
|
Invitrogen™
MA543645 |
100 μL |
Each for $522.50
|
|
|||||
Description
The pathological conditions of SRN1 correlate well with mutations at the NPHS2 gene, where expression of a protein known as Podocin occurs. Abnormal or inefficient signaling through Podocin protein-dependent networks contributes to the development of podocyte dysfunction and proteinuria. The human NPHS2 gene maps to chromosome 1q25-q31 and encodes a 383 amino acid protein. Podocin is an integral membrane protein that appears to fold into a hairpin-like structure with intracellular amino- and carboxy-termini. Transmembrane and cytoplasmic portions of Podocin share homology to the corresponding regions of the stomatin family proteins. Expression of high-order oligomers of Podocin in glomerular podocytes may reflect a scaffolding function that influences proper function of the glomerular filtration barrier, which is necessary for renal stability.Specifications
Podocin | |
1 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
AI790225; nephrosis 2 homolog, podocin; nephrosis 2 idiopathic steroid-resistant (podocin); nephrosis 2, idiopathic, steroid-resistant; nephrosis 2, idiopathic, steroid-resistant (podocin); nephrosis 2, podocin; NPHS2; NPHS2 podocin; NPHS2 stomatin family member, podocin; NPHS2, podocin; PDCN; podocin; SRN1 | |
Nphs2 | |
IgG2a κ | |
Protein A/G | |
Antibody |
C3 | |
Western Blot | |
Unconjugated | |
Mouse | |
Human, Mouse, Rat, Rat | |
Q8K4G9, Q91X05, Q9NP85 | |
170484, 170672, 7827 | |
Met222-Pro372 (Accession Q9NP85), with N-terminal His Tag. | |
Primary | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Nphs2 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title