Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Podocin/NPHS2 Rabbit anti-Human, Clone: 2O10G3, Novus Biologicals™
SDP

Catalog No. NB167313 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB167313 100 μg
NB167314 20 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB167313 Supplier Novus Biologicals Supplier No. NBP316111100UL
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Podocin/NPHS2 Monoclonal antibody specifically detects Podocin/NPHS2 in Human samples. It is validated for Western Blot

Specifications

Antigen Podocin/NPHS2
Applications Western Blot
Classification Monoclonal
Clone 2O10G3
Conjugate Unconjugated
Formulation PBS, 0.05% BSA, 50% glycerol, pH7.3
Gene Alias nephrosis 2, idiopathic, steroid-resistant (podocin), PDCNSRN1podocin
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 284-383 of human Podocin/NPHS2 (Q9NP85).
Purification Method Affinity purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 7827
Target Species Human
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.