Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Podocin/NPHS2 Rabbit anti-Human, Clone: 2O10G3, Novus Biologicals™

Rabbit Monoclonal Antibody
$205.50 - $458.50
Specifications
Antigen | Podocin/NPHS2 |
---|---|
Clone | 2O10G3 |
Applications | Western Blot |
Classification | Monoclonal |
Conjugate | Unconjugated |
Description
Podocin/NPHS2 Monoclonal antibody specifically detects Podocin/NPHS2 in Human samples. It is validated for Western BlotSpecifications
Podocin/NPHS2 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Signal Transduction | |
PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
7827 | |
IgG | |
Affinity purified |
2O10G3 | |
Monoclonal | |
Purified | |
RUO | |
Human | |
nephrosis 2, idiopathic, steroid-resistant (podocin), PDCNSRN1podocin | |
A synthetic peptide corresponding to a sequence within amino acids 284-383 of human Podocin/NPHS2 (Q9NP85). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title