Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Podoplanin Antibody (18H5) - Azide and BSA Free, Novus Biologicals™
SDP

Catalog No. NB018201 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.02 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB018201 0.02 mg
NB6001013 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB018201 Supplier Novus Biologicals Supplier No. NB60010130.02MG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody has been used in 2 publications

Podoplanin Monoclonal specifically detects Podoplanin in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, CyTOF-ready.

Specifications

Antigen Podoplanin
Applications CyTOF, Flow Cytometry, Western Blot, Immunohistochemistry, Immunofluorescence, Immunocytochemistry
Classification Monoclonal
Clone 18H5
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1-5 ug/ml, Flow Cytometry 5-10 ug/ml, Immunohistochemistry 6 - 30 ug/ml, Immunocytochemistry/Immunofluorescence 6-30 ug/ml, CyTOF-ready
Formulation PBS (pH 7.4) with No Preservative
Gene Accession No. Q86YL7
Gene Alias 36-KD, aggrus, Gp36, GP40, HT1A-1, hT1alpha-1, hT1alpha-2, lung type I cell membrane associated glycoprotein, lung type-I cell membrane-associated glycoprotein (T1A-2), OTS 8, OTS8, PA2.26 antigen, podoplanin, T1A, T1-alpha
Gene Symbols PDPN
Host Species Mouse
Immunogen MDCK cells stably expressing gp36 (Podoplanin) protein
Purification Method Protein G purified
Quantity 0.02 mg
Regulatory Status RUO
Research Discipline Core ESC Like Genes, Signal Transduction, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 10630
Test Specificity Human. Not yet tested in other species.
Reconstitution Centrifuge vial prior to opening. Reconstitute in sterile water to a concentration of > 0.5 mg/ml
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.