Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Podoplanin Antibody (8.1.1) - BSA Free, Novus Biologicals™

Golden Syrian Hamster Monoclonal Antibody has been used in 18 publications
Supplier: Novus Biologicals NB6001015SS
Description
Podoplanin Monoclonal specifically detects Podoplanin in Mouse, Human (Negative) samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Electron Microscopy, CyTOF-ready.Specifications
Podoplanin | |
Monoclonal | |
1 mg/mL | |
Western Blot 1:1000-1:2000, Flow Cytometry 1:400, Immunohistochemistry 1:100-1:500, Immunocytochemistry/Immunofluorescence 1-5ug/ml, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin 1:100-1:500, Immunohistochemistry-Frozen, Electron Microscopy, CyTOF-ready | |
36-KD, aggrus, Gp36, GP40, HT1A-1, hT1alpha-1, hT1alpha-2, lung type I cell membrane associated glycoprotein, lung type-I cell membrane-associated glycoprotein (T1A-2), OTS 8, OTS8, PA2.26 antigen, podoplanin, T1A, T1-alpha | |
Golden Syrian Hamster | |
40 kDa | |
0.025 mL | |
Core ESC Like Genes, Signal Transduction, Stem Cell Markers | |
10630 | |
Mouse, Human (Negative) | |
Purified |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation | |
8.1.1 | |
Unconjugated | |
Q62011 | |
PDPN | |
Murine thymic stromal cell lines | |
Protein G purified | |
RUO | |
Primary | |
Mouse. Does not cross-react with human. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction