Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Podoplanin Antibody (8.1.1) - BSA Free, Novus Biologicals™

Golden Syrian Hamster Monoclonal Antibody has been used in 18 publications
$207.00 - $487.50
Specifications
Antigen | Podoplanin |
---|---|
Clone | 8.1.1 |
Concentration | 1 mg/mL |
Dilution | Western Blot 1:1000-1:2000, Flow Cytometry 1:400, Immunohistochemistry 1:100-1:500, Immunocytochemistry/Immunofluorescence 1-5ug/ml, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin 1:100-1:500, Immunohistochemistry-Frozen, Electron Microscopy, CyTOF-ready |
Applications | Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB6001015SS
![]() |
Novus Biologicals
NB6001015SS |
0.025 mL |
Each for $207.00
|
|
|||||
NB6001015
![]() |
Novus Biologicals
NB6001015 |
0.1 mL |
Each for $487.50
|
|
|||||
Description
Podoplanin Monoclonal specifically detects Podoplanin in Mouse, Human (Negative) samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Electron Microscopy, CyTOF-ready.Specifications
Podoplanin | |
1 mg/mL | |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation | |
Unconjugated | |
Golden Syrian Hamster | |
Core ESC Like Genes, Signal Transduction, Stem Cell Markers | |
Q62011 | |
10630 | |
Murine thymic stromal cell lines | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
40 kDa |
8.1.1 | |
Western Blot 1:1000-1:2000, Flow Cytometry 1:400, Immunohistochemistry 1:100-1:500, Immunocytochemistry/Immunofluorescence 1-5ug/ml, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin 1:100-1:500, Immunohistochemistry-Frozen, Electron Microscopy, CyTOF-ready | |
Monoclonal | |
Purified | |
RUO | |
Mouse, Human (Negative) | |
36-KD, aggrus, Gp36, GP40, HT1A-1, hT1alpha-1, hT1alpha-2, lung type I cell membrane associated glycoprotein, lung type-I cell membrane-associated glycoprotein (T1A-2), OTS 8, OTS8, PA2.26 antigen, podoplanin, T1A, T1-alpha | |
PDPN | |
IgG | |
Protein G purified | |
Mouse. Does not cross-react with human. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title