Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Podoplanin Antibody (LF3/B7/D5B27), DyLight 755, Novus Biologicals™
SDP

Catalog No. NB11096423I Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB11096423I 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB11096423I Supplier Novus Biologicals Supplier No. NB11096423IR
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Podoplanin Monoclonal specifically detects Podoplanin in Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen Podoplanin
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone LF3/B7/D5B27, B7, D5B3
Conjugate DyLight 755
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
Gene Alias 36-KD, aggrus, Gp36, GP40, HT1A-1, hT1alpha-1, hT1alpha-2, lung type I cell membrane associated glycoprotein, lung type-I cell membrane-associated glycoprotein (T1A-2), OTS 8, OTS8, PA2.26 antigen, podoplanin, T1A, T1-alpha
Gene Symbols PDPN
Host Species Mouse
Immunogen Membrane protein fraction of isolated rat glomeruli
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Core ESC Like Genes, Signal Transduction, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 10630
Test Specificity Human (Negative), Mouse (Negative).
Target Species Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.