Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Podoplanin Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB60010140.025MG
Description
Podoplanin Polyclonal specifically detects Podoplanin in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, CyTOF-ready.Specifications
Podoplanin | |
Polyclonal | |
Unconjugated | |
PBS with No Preservative | |
PDPN | |
Recombinant human Podoplanin | |
0.025 mg | |
Core ESC Like Genes, Signal Transduction, Stem Cell Markers | |
10630 | |
Reconstitute in sterile water to a concentration of 0.1-1.0mg/mL. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
IgG |
CyTOF, Western Blot, Immunocytochemistry, Immunoprecipitation, Immunofluorescence, Flow Cytometry | |
LYOPH | |
Western Blot 2-4 ug/mL, Flow Cytometry 1:10-1:1000, Immunocytochemistry/Immunofluorescence 10 ug/mL, Immunoprecipitation 1:10-1:500, CyTOF-ready | |
36-KD, aggrus, Gp36, GP40, HT1A-1, hT1alpha-1, hT1alpha-2, lung type I cell membrane associated glycoprotein, lung type-I cell membrane-associated glycoprotein (T1A-2), OTS 8, OTS8, PA2.26 antigen, podoplanin, T1A, T1-alpha | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Podoplanin | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction