Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Podoplanin Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$206.50 - $501.50
Specifications
Antigen | Podoplanin |
---|---|
Concentration | LYOPH |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Purified |
Description
Podoplanin Polyclonal specifically detects Podoplanin in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, CyTOF-ready.Specifications
Podoplanin | |
Polyclonal | |
Purified | |
RUO | |
Human | |
36-KD, aggrus, Gp36, GP40, HT1A-1, hT1alpha-1, hT1alpha-2, lung type I cell membrane associated glycoprotein, lung type-I cell membrane-associated glycoprotein (T1A-2), OTS 8, OTS8, PA2.26 antigen, podoplanin, T1A, T1-alpha | |
PDPN | |
IgG | |
Protein A purified | |
Podoplanin |
LYOPH | |
Unconjugated | |
Rabbit | |
Core ESC Like Genes, Signal Transduction, Stem Cell Markers | |
PBS with No Preservative | |
10630 | |
Recombinant human Podoplanin | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title