Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Podoplanin Antibody (PMab-1), Alexa Fluor™ 405, Novus Biologicals™
SDP

Catalog No. NB152004 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB152004 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB152004 Supplier Novus Biologicals Supplier No. NBP311971AF405
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Podoplanin Monoclonal antibody specifically detects Podoplanin in Mouse samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry

Specifications

Antigen Podoplanin
Applications Western Blot, Flow Cytometry, ELISA, Immunohistochemistry
Classification Monoclonal
Clone PMab-1
Conjugate Alexa Fluor 405
Dilution Western Blot, Flow Cytometry, ELISA, Immunohistochemistry
Formulation 50mM Sodium Borate
Gene Alias 36-KD, aggrus, Gp36, GP40, HT1A-1, hT1alpha-1, hT1alpha-2, lung type I cell membrane associated glycoprotein, lung type-I cell membrane-associated glycoprotein (T1A-2), OTS 8, OTS8, PA2.26 antigen, podoplanin, T1A, T1-alpha
Host Species Rabbit
Immunogen This antibody was raised by immunising rats with 14-residue synthetic peptide mpp3851, which corresponds to amino acids 38-51 (GDGMVPPGIEDKIT) of the platelet-aggregation-stimulating (PLAG) domain of mouse podoplanin. Spleen cells were then harvested, and fused to P3U1 cells to generate stable hybridomas.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Core ESC Like Genes, Signal Transduction, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 10630
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Isotype IgG κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.