Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Podoplanin Antibody (pmab-1) - Azide and BSA Free, Novus Biologicals™
SDP

Catalog No. NB013990 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB013990 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB013990 Supplier Novus Biologicals Supplier No. NBP280917
Add to Cart
Add to Cart

Rat Monoclonal Antibody

Podoplanin Monoclonal specifically detects Podoplanin in Mouse samples. It is validated for Western Blot, Flow Cytometry, CyTOF-ready.

Specifications

Antigen Podoplanin
Applications Western Blot, Flow Cytometry, CyTOF
Classification Monoclonal
Clone pmab-1
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 3 - 5 μg/mL, Flow Cytometry : 0.5 ug/10^5 cells, CyTOF-ready
Formulation PBS
Gene Alias 36-KD, aggrus, Gp36, GP40, HT1A-1, hT1alpha-1, hT1alpha-2, lung type I cell membrane associated glycoprotein, lung type-I cell membrane-associated glycoprotein (T1A-2), OTS 8, OTS8, PA2.26 antigen, podoplanin, T1A, T1-alpha
Gene Symbols PDPN
Host Species Rat
Immunogen Podoplanin
Purification Method Protein G purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Core ESC Like Genes, Signal Transduction, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 10630
Target Species Mouse
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.