Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Podoplanin Antibody (pmab-1), DyLight 650, Novus Biologicals™
SDP

Catalog No. NBP203955C Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP203955C 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP203955C Supplier Novus Biologicals Supplier No. NBP203955C
Add to Cart
Add to Cart

Rat Monoclonal Antibody

Podoplanin Monoclonal specifically detects Podoplanin in Mouse samples. It is validated for Western Blot, Flow Cytometry.

Specifications

Antigen Podoplanin
Applications Western Blot, Flow Cytometry
Classification Monoclonal
Clone pmab-1
Conjugate DyLight 650
Dilution Western Blot, Flow Cytometry
Gene Alias 36-KD, aggrus, Gp36, GP40, HT1A-1, hT1alpha-1, hT1alpha-2, lung type I cell membrane associated glycoprotein, lung type-I cell membrane-associated glycoprotein (T1A-2), OTS 8, OTS8, PA2.26 antigen, podoplanin, T1A, T1-alpha
Gene Symbols PDPN
Host Species Rat
Immunogen The immunogen is Podoplanin.
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Core ESC Like Genes, Signal Transduction, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 10630
Target Species Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2a κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.