Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Podoplanin Antibody (RTD4E10) - Azide and BSA Free, Novus Biologicals™
SDP

Catalog No. NB019659 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.02 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB019659 0.02 mg
NBP236058 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB019659 Supplier Novus Biologicals Supplier No. NBP2360580.02MG
Add to Cart
Add to Cart

Hamster Monoclonal Antibody

Podoplanin Monoclonal specifically detects Podoplanin in Mouse, Human (Negative) samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin.

Specifications

Antigen Podoplanin
Applications ELISA, Immunoprecipitation, Western Blot, Immunofluorescence, Immunohistochemistry (Paraffin), Immunocytochemistry, Immunohistochemistry
Classification Monoclonal
Clone RTD4E10
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1-5ug/ml, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1-10 ug/ml, Immunoprecipitation 2-10 ug/ml, Immunohistochemistry-Paraffin 1-10 ug/ml
Formulation PBS with No Preservative
Gene Alias 36-KD, aggrus, Gp36, GP40, HT1A-1, hT1alpha-1, hT1alpha-2, lung type I cell membrane associated glycoprotein, lung type-I cell membrane-associated glycoprotein (T1A-2), OTS 8, OTS8, PA2.26 antigen, podoplanin, T1A, T1-alpha
Gene Symbols PDPN
Host Species Hamster
Immunogen recombinant mouse protein of IL-13
Purification Method Protein G purified
Quantity 0.02 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10630
Reconstitution Reconstitute the antibody with 500 ul sterile PBS and the final concentration is 200 ug/ml.
Target Species Mouse
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.