Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Podoplanin Mouse anti-Human, PE, Clone: 56F7, Novus Biologicals™
SDP

Catalog No. NB161486 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB161486 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB161486 Supplier Novus Biologicals Supplier No. NBP259445PE
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Podoplanin Monoclonal antibody specifically detects Podoplanin in Human samples. It is validated for Flow Cytometry

Specifications

Antigen Podoplanin
Applications Flow Cytometry
Classification Monoclonal
Clone 56F7
Conjugate PE
Formulation PBS
Gene Alias 36-KD, aggrus, Gp36, GP40, HT1A-1, hT1alpha-1, hT1alpha-2, lung type I cell membrane associated glycoprotein, lung type-I cell membrane-associated glycoprotein (T1A-2), OTS 8, OTS8, PA2.26 antigen, podoplanin, T1A, T1-alpha
Host Species Mouse
Immunogen Recombinant human Podoplanin (99-207aa) purified from E. coli
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Core ESC Like Genes, Signal Transduction, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 10630
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.