Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

POGK Antibody (OTI6D5), DyLight 755, Novus Biologicals™
SDP

Catalog No. NBP273513IR Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP273513IR 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP273513IR Supplier Novus Biologicals Supplier No. NBP273513IR
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

POGK Monoclonal specifically detects POGK in Human, Mouse, Rat, Monkey samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen POGK
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone OTI6D5
Conjugate DyLight 755
Dilution Western Blot, Immunohistochemistry
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias BASS2, KIAA1513, KIAA15131, KRAB box domain containing 2, KRBOX2, LST003, pogo transposable element with KRAB domain
Gene Symbols POGK
Host Species Mouse
Immunogen Full length recombinant protein of human POGK (NP_060012) produced in HEK293T cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Apoptosis
Primary or Secondary Primary
Gene ID (Entrez) 57645.0
Target Species Human, Mouse, Rat, Monkey
Content And Storage Store at 4°C in the dark.
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.