Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

POGZ Antibody (HL1898) - Azide and BSA Free, Novus Biologicals™
SDP

Catalog No. NB295009 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB295009 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB295009 Supplier Novus Biologicals Supplier No. NBP325608
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

POGZ Monoclonal antibody specifically detects POGZ in Human samples. It is validated for Western Blot

Specifications

Antigen POGZ
Applications Western Blot
Classification Monoclonal
Clone HL1898
Conjugate Unconjugated
Dilution Western Blot 1:500-1:3000
Formulation PBS
Gene Alias KIAA0461ZNF635, pogo transposable element with ZNF domain, putative protein product of Nbla00003, SUHW5MGC71543, Suppressor of hairy wing homolog 5, Zinc finger protein 280E, Zinc finger protein 635, ZNF280EZNF635m
Host Species Rabbit
Immunogen Synthetic peptide of human POGZ.
Purification Method Protein A purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23126
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.