Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ POGZ Polyclonal Antibody
GREENER_CHOICE

Catalog No. pipa552349
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA552349 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA552349 Supplier Invitrogen™ Supplier No. PA552349
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: GEPWCDVVLA ILADGTVLPT LVFYRGQMDQ PANMPDSILL EAKESGYSDD EIMELWSTRV WQKHTACQRS KGMLVMDCHR THLSEEVLAM LSASSTLPAV VPAGCSSKIQ PLDVCIKRTV KNFLH Highest antigen sequence identity to the following orthologs: Mouse - 91%, Rat - 91%.

POGZ appears to be a zinc finger protein containing a transposase domain at the C-terminus. This protein was found to interact with the transcription factor SP1 in a yeast two-hybrid systemThe protein encoded by this gene appears to be a zinc finger protein containing a transposase domain at the C-terminus. This protein was found to interact with the transcription factor SP1 in a yeast two-hybrid system. At least three alternatively spliced transcript variants encoding distinct isoforms have been observed.
TRUSTED_SUSTAINABILITY

Specifications

Antigen POGZ
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene POGZ
Gene Accession No. Q7Z3K3
Gene Alias 9530006B08Rik; KIAA0461; MRD37; Nbla00003; pogo transposable element derived with ZNF domain; pogo transposable element with ZNF domain; POGZ; putative protein product of Nbla00003; SUHW5; Suppressor of hairy wing homolog 5; WHSUS; Zinc finger protein 280E; zinc finger protein 635; ZNF280E; ZNF635; ZNF635m
Gene Symbols POGZ
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human POGZ. Recombinant protein control fragment (Product #RP-89485).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23126
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.