Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Pointe Scientific Albumin Reagents
Click to view available options
Quantity:
15 mL
120 mL
500 mL
1000 mL
Type:
Albumin Reagent Kit
Albumin Standard Reagent Kit
Description
Reagent requires Albumin Standard (23-666-066)/Chemistry Calibrator (23-666-189). HA901-1400 is used for Hitachi 911, 912, 914. HA701-600 is used for Hitachi 704. HA401-3000 is used for Hitachi 747.
Specifications
Specifications
Certifications/Compliance | CE Marked |
Type | Albumin Reagent Kit |
Formulation | Bromocresol green 0.15g/L, Buffer pH 4.6 ±0.1, Surfactants, non-reactive ingredients and Stabilizers, Sodium Azide 0.01% |
For Use With (Application) | Quantitative determination of Albumin in Serum, in-vitro diagnostic use only |
pH | 4.66 |
Quantity | 1000 mL |
Form | Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction