Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pointe Scientific ALT (SGPT) Liquid Reagents

Catalog No. p-318166
Click to view available options
Quantity:
1 x 120mL R1, 1 x 30mL R2
1 x 500mL R1, 1 x 125mL R2
3 x 120mL R1, 1 x 90mL R2
This item is not returnable. View return policy
This item is not returnable. View return policy

For quantitative determination of Alanine Aminotransferase in serum

Reagent Set 1 includes 1 x 120mL substrate/enzyme reagent and 1 x 30mL coenzyme reagent.
Reagent Set 2 includes 3 x 120mL substrate/enzyme reagent and 1 x 90mL coenzyme reagent.
Reagent Set 3 includes 1 x 500mL substrate/enzyme reagent and 1 x 125mL coenzyme reagent.

  • Method: Modified IFCC, Kinetic
  • Format: Liquid
  • Wavelength: 340nm
  • Linearity: 0-500 U/L
  • Expected Values: 4-36 U/L
  • Storage Temperature: 2° to 8°C
  • Reconstituted Stability: Ready to use liquid

Specifications

Certifications/Compliance CE Marked
Type Alanine Aminotransferase Reagent
Formulation L-alanine 500mM, alpha-Ketoglutaric Acid 15mM, LDH(microbial) >2000IU/L, NADH >0.18mM, Buffer 100mM pH 7.5 ±0.1, Sodium Azide 0.25%, Stabilizers
For Use With (Application) Quantitative determination of Alanine Aminotransferase in Serum
Quantity 1 x 120mL R1, 1 x 30mL R2
Form Liquid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.