Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pointe Scientific Ammonia Reagents

Catalog No. p-318397
Click to view available options
Quantity:
10 x 6.5mL Substrate, 10 x 2mL Enzyme
4 x 6.5mL Substrate, 4 x 2mL Enzyme
Formulation:
Ammonia Enzyme/R2: Buffer 100mM/L, EDTA 2mM/L, Adenosine Diphosphate 0.5mM/L, GLDH 400 Kμ/L, fillers and Stabilizers, pH 7.8 ± 0.1
Ammonia Substrate/R1: Buffer 100mM/L, EDTA 2mM/L, alpha-Ketoglutarate 3.4mM/L, Adenosine Diphosphate 0.5mM/L, NADPH 0.3mM/L, fillers and Stabilizers, pH 8.6 ± 0.1
Includes:
Reagent set 1 includes 4 x 6.5mL substrate reagent and 4 x 2mL of Enzyme reagent
Reagent set 2 includes 10 x 6.5mL substrate reagent and 10 x 2mL of Enzyme reagent
This item is not returnable. View return policy
This item is not returnable. View return policy

Reagent Set 1 includes 4 x 6.5mL substrate reagent and 4 x 2mL of enzyme reagent.
Reagent Set 2 includes 10 x 6.5mL substrate reagent and 10 x 2mL of enzyme reagent.

Specifications

Certifications/Compliance CE Marked
Type Ammonia Reagent
Formulation Ammonia Substrate/R1: Buffer 100mM/L, EDTA 2mM/L, alpha-Ketoglutarate 3.4mM/L, Adenosine Diphosphate 0.5mM/L, NADPH 0.3mM/L, fillers and Stabilizers, pH 8.6 ± 0.1
For Use With (Application) Enzymatic measurement of Ammonia in Plasma, in-vitro diagnostic use only
Includes Reagent set 1 includes 4 x 6.5mL substrate reagent and 4 x 2mL of Enzyme reagent
Quantity 4 x 6.5mL Substrate, 4 x 2mL Enzyme
Form Dry Powder
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.