Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Pointe Scientific Ammonia Reagents
Shop All Pointe Scientific Products
Click to view available options
Quantity:
10 x 6.5mL Substrate, 10 x 2mL Enzyme
4 x 6.5mL Substrate, 4 x 2mL Enzyme
Formulation:
Ammonia Enzyme/R2: Buffer 100mM/L, EDTA 2mM/L, Adenosine Diphosphate 0.5mM/L, GLDH 400 Kμ/L, fillers and Stabilizers, pH 7.8 ± 0.1
Ammonia Substrate/R1: Buffer 100mM/L, EDTA 2mM/L, alpha-Ketoglutarate 3.4mM/L, Adenosine Diphosphate 0.5mM/L, NADPH 0.3mM/L, fillers and Stabilizers, pH 8.6 ± 0.1
Includes:
Reagent set 1 includes 4 x 6.5mL substrate reagent and 4 x 2mL of Enzyme reagent
Reagent set 2 includes 10 x 6.5mL substrate reagent and 10 x 2mL of Enzyme reagent
Description
Reagent Set 1 includes 4 x 6.5mL substrate reagent and 4 x 2mL of enzyme reagent.
Reagent Set 2 includes 10 x 6.5mL substrate reagent and 10 x 2mL of enzyme reagent.
Specifications
Specifications
| Certifications/Compliance | CE Marked |
| Type | Ammonia Reagent |
| Formulation | Ammonia Enzyme/R2: Buffer 100mM/L, EDTA 2mM/L, Adenosine Diphosphate 0.5mM/L, GLDH 400 Kμ/L, fillers and Stabilizers, pH 7.8 ± 0.1 |
| For Use With (Application) | Enzymatic measurement of Ammonia in Plasma, in-vitro diagnostic use only |
| Includes | Reagent set 2 includes 10 x 6.5mL substrate reagent and 10 x 2mL of Enzyme reagent |
| Quantity | 10 x 6.5mL Substrate, 10 x 2mL Enzyme |
| Form | Dry Powder |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction