Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pointe Scientific Amylase-EPS Reagents

Catalog No. p-2715671 Shop All Pointe Scientific Products
Click to view available options
Quantity:
3 x 120 R2mL
3 x 270 R1mL
6 x 22mL R1, 3 x 10mL R2
Formulation:
Buffer 50mM pH 7.2 ±0.1, NaCl >50mM, MgCl2 10mM, alpha-Glucosidase >2KU/L, EPS-G7 >1mM
Sodium Azide 0.095%
For Use With (Equipment):
Hitachi 704 analyzers
Hitachi 747 analyzers
Type:
Amylase-EPS R1
Amylase-EPS R2
Amylase-EPS Reagent Kit

HA965-324 is for Hitachi 911, 912, 914. HA765-162 is for Hitachi 704. HA465-R1 and HA465-R2 are for Hitachi 747.

Specifications

Certifications/Compliance CE Marked
Type Amylase-EPS R1
Formulation Sodium Azide 0.095%
For Use With (Application) Quantitative kinetic determination of a-amylase activity in Human Serum
For Use With (Equipment) Hitachi 747 analyzers
Quantity 3 x 270 R1mL
Form Liquid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.