Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Pointe Scientific AST (SGOT) Liquid Reagents
Click to view available options
Quantity:
1 x 120 mL R1, 1 x 30 mL R2
1 x 500 mL R1, 1 x 125 mL R2
3 x 120 mL R1, 1 x 90 mL R2
Description
Set 1 consists of 1 x 120mL substrate reagent and 1 x 30mL enzyme reagent.
Set 2 consists of 3 x 120mL substrate reagent and 1 x 90mL enzyme reagent.
Set 3 consists of 1 x 500mL substrate reagent and 1 x 125mL enzyme reagent.
- Method: Modified IFCC, Kinetic
- Format: Liquid
- Linearity: 0-500 U/L
- Expected Values: 4-36 U/L
- Reconstituted Stability: Ready to use liquid
Specifications
Specifications
Certifications/Compliance | CE Marked |
Type | Aspartate Aminotransferase Reagent Kit |
Formulation | L-aspartic Acid >200mM, alpha-Ketoglutaric Acid 12mM, LDH (microbial) >1000U/L, MDH (microbial) >800U/L, NADH >0.18mM, pH 7.8 ±0.1 Buffer, Sodium Azide 0.25%, Stabilizers |
For Use With (Application) | Quantitative determination of Aspartate Aminotransferase in Human Serum |
Quantity | 3 x 120 mL R1, 1 x 90 mL R2 |
Form | Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction