Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Pointe Scientific Auto LDL Cholesterol Liquid Reagents
Click to view available options
Quantity:
1 x 240mL R1, 1 x 80mL R2
1 x 30mL R1, 1 x 10mL R2
1 x 60mL R1, 1 x 20mL R2
Type:
AutoLDL™ Cholesterol Reagent
AutoLDL™ Cholesterol Reagent Kit
Description
Requires autoHDL/LDL Calibrator (23-666-140).
- Method: Direct/Automated
- Format: Liquid
- Wavelength: 546/660nm
- Linearity: 0-700mg/dL
- Expected Values: >130mg/dL
- Reconstituted Stability: Ready to use liquid
Specifications
Specifications
Certifications/Compliance | CE Marked |
Type | AutoLDL™ Cholesterol Reagent |
Formulation | R1 contains: MES Buffer pH 6.3, Detergent 1, Cholesterol Esterase, Cholesterol Oxidase, Peroxidase, 4-aminoantipyrine, Ascorbic Acid Oxidase, Preservative, R2 contains: MES Buffer pH 6.3, Detergent 2, N,N-bis (4-sulfhobutyl)-m Toluidine-disodium (DSBmT), |
For Use With (Application) | Quantitative determination of Low Density Lipoprotein Cholesterol in Human Serum or Plasma |
Quantity | 1 x 240mL R1, 1 x 80mL R2 |
Form | Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction