Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Pointe Scientific autoHDL Cholesterol Liquid Reagents
Click to view available options
Quantity:
1 x 240L R1, 1 x 800mL R2
1 x 30mL R1, 1 x 10mL R2
1 x 60mL R1, 1 x 20mL R2
1 x 750mL R1, 1 x 250mL R2
Description
Reagent requires autoHDL/LDL Calibrator (23-666-140).
- Method: Direct/Automated
- Format: Liquid
- Wavelength: 600/70nm
- Linearity: 2 - 150mg/dL
- Expected Values: 30-85mg/dL
- Storage Temperature: 2° to 8°C
- Reconstituted Stability: Ready to use liquid
Specifications
Specifications
Certifications/Compliance | CE Marked |
Type | AutoHDL™ Cholesterol Liquid Reagent Kit |
For Use With (Application) | Quantitative determination high-density Lipoprotein Cholesterol in Human Serum or Plasma |
Quantity | 1 x 750mL R1, 1 x 250mL R2 |
Form | Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction