Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Pointe Scientific Bilirubin Total Reagents for Hitachi Analyzers
Shop All Pointe Scientific Products
Click to view available options
Quantity:
5 x 175 R2mL
5 x 600 R1mL
6 x 45mL R1, 3 x 25mL R2
Formulation:
Acid Buffer >30mM/L, DPD >2mM/L and Stabilizers
R1 contains: Acid Buffer 50mM/L, Surfactant, R2 contains: Acid Buffer >30mM/L, DPD >2mM/L and Stabilizers
Total Bilirubin R1 Acid Buffer 50mM/L, Surfactants, Total Bilirubin R2 Acid Buffer >30mM/L, DPD >2mM/L and Stabilizers
Specifications
Specifications
| Certifications/Compliance | CE Marked |
| Type | Total Bilirubin Reagent Kit |
| Formulation | Total Bilirubin R1 Acid Buffer 50mM/L, Surfactants, Total Bilirubin R2 Acid Buffer >30mM/L, DPD >2mM/L and Stabilizers |
| For Use With (Application) | Quantitative determination of total Bilirubin in Serum, in-vitro diagnostic use only |
| Quantity | 5 x 600 R1mL |
| Form | Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction