Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Pointe Scientific Calcium Liquid Reagents
Shop All Pointe Scientific Products
Click to view available options
Quantity:
2 x 120mL Color, 2 x 120mL Buffer
2 x 30mL Color, 2 x 30mL Buffer
Description
Includes
- Color Reagent: o-cresolphthalein complexone 0.11mM, 8-hydroxyquinoline 17.0mM, surfactant
- Buffer Reagent: 2-Amino-2-Methyl-1-Propanol 976mM, Potassium Cyanide 2.0mM
- Method: Cresolphthalein Complexone (CPC), Endpoint
- Format: Liquid (ready to use)
- Wavelength: 570nm
- Linearity: 20mg/dL
- Expected Values: 8.5 to 10.4mg/dL
- Separate color and buffer reagents
For quantitative determination of calcium in serum
Specifications
Specifications
| Certifications/Compliance | CE Marked |
| Type | Calcium Reagent Kit |
| For Use With (Application) | Quantitative determination of Calcium in Serum |
| Includes | Color reagent : o-Cresolphthalein Complexone 0.11mM, 8-Hydroxyquinoline 17.0mM, Surfactant, Buffer reagent : 2-Amino-2-Methyl-1-Propanol 976mM, 2mM Potassium Cyanide |
| Quantity | 2 x 120mL Color, 2 x 120mL Buffer |
| Form | Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction