Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pointe Scientific Calcium Reagents for Hitachi™ Analyzers

Catalog No. p-2717127
Click to view available options
Quantity:
5 x 300 mL R2
5 x 600 mL R1
7 x 100 mL R1, 3 x 100 mL R2
7 x 50 mL R1, 3 x 50 mL R2
For Use With (Equipment):
Hitachi 704, 902 Analyzers
Hitachi 717, 911, 912, 914 Analyzers
Hitachi 747 Analyzers

Includes

  • Reagent 1: 2-Amino-2-Methyl-1-Propanol, pH 10.6±0.1
  • Reagent 2: o-CPC 0.3mM, surfactant

Quantify calcium levels easily with these instrument-specific reagents

  • Method: Cresolphthalein Complexone (CPC)
  • Format: Liquid (ready to use)
  • Linearity: 20mg/dL
  • Expected Values: 8.5 to 10.5mg/dL

Applications:

For quantitative determination of calcium in serum on Hitachi analyzers

Specifications

Certifications/Compliance CE Marked
Type Calcium Reagent Kit
Formulation 2-Amino-2-Methyl-1-Propanol, pH 10.6 ±0.1
For Use With (Application) Quantitative determination of Calcium in Serum
For Use With (Equipment) Hitachi 747 Analyzers
Quantity 5 x 600 mL R1
Form Liquid

For in vitro diagnostic use only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.