Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Pointe Scientific CK (Liquid) Reagents
Shop All Pointe Scientific Products
Click to view available options
Quantity:
150 mL
450 mL
625 mL
Specifications
Specifications
| Type | Creatine Kinase Reagent Kit |
| Formulation | CK R1 (Buffer) contains: Imidazole Buffer 100mM/L (pH 6.7), NADP 2mM/L, HK (Baker's Yeast) 2.5KU/L, Glucose 20mM/L, Magnesium Acetate 10mM/L, EDTA 2mM/L, N-acetylcysteine (NAC) 20mM/L, CK R2 (enzyme reagent) contains: Imidazole Buffer 100mM/L (pH 6.7), A |
| For Use With (Application) | Quantitative determination of Creatine kinase activity in Serum |
| Quantity | 625 mL |
| Form | Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction