Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Pointe Scientific COBAS MIRA™ Reagent Containers and Sample Cups
Shop All Pointe Scientific Products
Click to view available options
Volume (Metric):
4 mL
10 mL
Product Type:
Reagent Container
Sample Cup
Description
Sizes for Reagent Containers: 4mL (0.14 oz.) and 10 mL (0.34 oz.)
Specifications
Specifications
| Product Type | Reagent Container |
| Volume (Metric) | 4 mL |
| For Use With (Equipment) | Cobas Mira™ analyzer |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction