Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pointe Scientific Creatinine Reagents for Hitachi™ Analyzers

Catalog No. 23666232 Shop All Pointe Scientific Products
Click to view available options
Quantity:
11 x 100mL R1, 3 x 80mL R2
5 x 300 R2mL
5 x 600 R1mL
8 x 50mL R1, 2 x 50mL R2
Formulation:
Creatinine R1 contains 0.68% Lithium Hydroxide, Alkaline Buffer
Creatinine R1 contains 0.68% Lithium Hydroxide, Alkaline Buffer, Creatinine R2 contains Picric Acid (dilute) 0.9%, Lithium Hydroxide (dilute) 0.17%
Creatinine R2 contains Picric Acid (dilute) 0.9%, Lithium Hydroxide (dilute) 0.17%

Includes

  • Reagent 1: Alkaline buffer
  • Reagent 2: Picric Acid 40mM, surfactant

Quantify creatinine easily with system specific reagents

  • Method: Modified Jaffe, Initial Rate
  • Format: Liquid (ready to use)
  • Linearity: 25.0mg/dL
  • Expected Values: 0.40 to 1.40mg/dL

Specifications

Certifications/Compliance CE Marked
Type Creatinine Reagent Kit
Formulation Creatinine R1 contains 0.68% Lithium Hydroxide, Alkaline Buffer, Creatinine R2 contains Picric Acid (dilute) 0.9%, Lithium Hydroxide (dilute) 0.17%
For Use With (Application) Quantitative determination of Creatinine in Serum
Quantity 8 x 50mL R1, 2 x 50mL R2
Form Liquid

For in vitro diagnostic use only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.